Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052979.1 | NP_443211.1 | 4 | 436 | ||
GenBank mRNA | AB045587.1 | - | 4 | 436 |