Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012599.2 | NP_036731.2 | 4, 5 | 411 | ||
GenBank mRNA | BC088159.1 | - | 4, 5 | 411 | 7326244 | |
FQ209936.1 | - | 4, 5 | 427 | |||
FQ218582.1 | - | 4, 5 | 439 |