Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012614.2 | NP_036746.1 | 2 | 143 | ||
GenBank mRNA | M15880.1 | - | 2 | 118 | ||
M20373.1 | - | 2 | 98 |
Gene Symbol: LOC100912228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749745.3 | XP_003749793.1 | 156 |