Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271086.1 | NP_001258015.1 | 2 | 444 | ||
XM_006230213.2 | XP_006230275.1 | 2 | 683 | |||
XM_008759858.1 | XP_008758080.1 | 2 | 1021 | |||
GenBank mRNA | Y00090.1 | - | 1 | 307 |