Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012904.2 | NP_037036.1 | 8 | 652 | ||
XM_006231159.1 | XP_006231221.1 | 9 | 820 | |||
XM_008760295.1 | XP_008758517.1 | 8 | 739 | |||
GenBank mRNA | BC061710.1 | - | 8 | 650 | 5598112 | |
FQ213489.1 | - | 8 | 667 | |||
FQ215033.1 | - | 8 | 667 | |||
FQ216134.1 | - | 8 | 667 | |||
FQ228716.1 | - | 8 | 668 | |||
FQ229071.1 | - | 8 | 668 | |||
FQ229198.1 | - | 7 | 588 | |||
M19967.1 | - | 7 | 619 | |||
Y00446.1 | - | 8 | 653 |