Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013192.2 | NP_037324.1 | 2 | 298 | ||
XM_008768567.1 | XP_008766789.1 | 1 | 84 | |||
GenBank mRNA | AB073753.1 | - | 2 | 298 | ||
AB073754.1 | - | 2 | 298 | |||
AB073755.1 | - | 2 | 298 | |||
AB073756.1 | - | 2 | 298 | |||
AY095171.1 | - | 1 | 87 | |||
U21087.1 | - | 2 | 298 |