Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bles03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024233.2 | NP_001019404.2 | 2 | 413 | ||
NM_001243534.1 | NP_001230463.1 | 2 | 427 | |||
GenBank mRNA | BC093387.1 | - | 2 | 272 | 7389419 | |
CK483724.1 | - | 2 | 269 | |||
CK596744.1 | - | 2 | 413 | |||
FQ233132.1 | - | 2 | 306 |