Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iscu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105936.1 | NP_001099406.1 | 3 | 323 | ||
GenBank mRNA | BC167056.1 | - | 3 | 302 | 7369214 |