Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ormdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105940.1 | NP_001099410.1 | 3 | 440 | ||
XM_006240753.2 | XP_006240815.1 | 3 | 420 | |||
XM_006240754.2 | XP_006240816.1 | 3 | 340 |