Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106010.1 | NP_001099480.1 | 1 | 663 | ||
XM_006251045.2 | XP_006251107.1 | 3 | 1062 | |||
XM_006251046.2 | XP_006251108.1 | 4 | 1165 | |||
XM_006251047.2 | XP_006251109.1 | 3 | 1063 | |||
XM_008770205.1 | XP_008768427.1 | 3 | 1063 | |||
GenBank mRNA | BC168903.1 | - | 2 | 841 |