Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106049.1 | NP_001099519.1 | 2 | 393 | ||
XM_006252294.2 | XP_006252356.1 | 1 | 708 | |||
XM_008770862.1 | XP_008769084.1 | 2 | 496 |