Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006968.1 | NP_001006969.1 | 756 | |||
XM_006228017.2 | XP_006228079.1 | 1190 | ||||
XM_006228018.2 | XP_006228080.1 | 764 | ||||
XM_008758752.1 | XP_008756974.1 | 1369 | ||||
GenBank mRNA | BC083767.1 | - | 756 | 7193201 |
Gene Symbol: LOC100909510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748716.3 | XP_003748764.1 | 4 | 1126 | ||
XM_003748717.3 | XP_003748765.1 | 4 | 764 | |||
XM_006228075.2 | XP_006228137.1 | 4 | 1190 | |||
XM_008758839.1 | XP_008757061.1 | 3 | 1369 |