Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106317.2 | NP_001099787.1 | 2 | 235 | ||
XM_006230537.2 | XP_006230599.1 | 3 | 335 | |||
XM_006230538.2 | XP_006230600.1 | 3 | 628 | |||
XM_006230539.2 | XP_006230601.1 | 2 | 231 | |||
GenBank mRNA | BC158743.1 | - | 2 | 235 | 9100666 |