Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psbpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207593.2 | NP_997476.2 | 3 | 313 | ||
GenBank mRNA | BC099748.1 | - | 3 | 307 |