Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106339.1 | NP_001099809.1 | 2 | 148 | ||
XM_006231087.2 | XP_006231149.1 | 3 | 317 | |||
XM_006231088.2 | XP_006231150.1 | 3 | 317 |
Gene Symbol: LOC690862 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760276.1 | XP_008758498.1 | 3 | 313 |