Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134959.1 | NP_001128431.1 | 9, 10 | 1029 | ||
XM_008761138.1 | XP_008759360.1 | 9, 10 | 1237 | |||
XM_008761139.1 | XP_008759361.1 | 7, 8 | 1067 | |||
XM_008761140.1 | XP_008759362.1 | 9, 10 | 1236 | |||
GenBank mRNA | BC168665.1 | - | 9, 10 | 1029 | 7933243 | |
FQ226108.1 | - | 9, 10 | 1075 | |||
FQ231267.1 | - | 9, 10 | 1075 |