Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011952.1 | NP_001011952.1 | 5 | 958 | ||
XM_006233356.2 | XP_006233418.1 | 6 | 997 | |||
XM_006233357.2 | XP_006233419.1 | 6 | 1217 | |||
GenBank mRNA | BC089844.1 | - | 5 | 958 | 7368889 |