Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168549.1 | NP_001162021.1 | 6, 7 | 1191 | ||
XM_006233672.2 | XP_006233734.1 | 6, 7 | 1159 | |||
XM_006233673.2 | XP_006233735.1 | 5, 6 | 1126 | |||
XM_006233674.2 | XP_006233736.1 | 5, 6 | 1126 | |||
XM_006233675.2 | XP_006233737.1 | 6, 7 | 1159 |