Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047867.1 | NP_001041332.1 | 11 | 1125 | ||
GenBank mRNA | AY325240.1 | - | 11 | 1125 |