Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Magoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100536.1 | NP_001094006.1 | 5 | 482 | ||
XM_006238510.2 | XP_006238572.1 | 5 | 574 | |||
GenBank mRNA | BQ196337.1 | - | 1 | 185 | ||
CK845348.1 | - | 1 | 151 | |||
DQ359101.1 | - | 5 | 411 |