Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100538.1 | NP_001094008.1 | 8 | 1549 | ||
XM_006238798.1 | XP_006238860.1 | 8 | 2089 | |||
XM_006238799.1 | XP_006238861.1 | 8 | 2089 | |||
XM_006238800.1 | XP_006238862.1 | 8 | 2089 | |||
XM_006238801.1 | XP_006238863.1 | 8 | 2089 | |||
GenBank mRNA | BC088337.1 | - | 3 | 448 | ||
BC166452.1 | - | 8 | 1554 | 8372968 |
Gene Symbol: RGD1562451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769025.1 | XP_008767247.1 | 1 | 1523 |