Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127525.1 | NP_001120997.1 | 3 | 299 | ||
GenBank mRNA | BC161895.1 | - | 3 | 299 | 7304509 | |
FQ213771.1 | - | 3 | 287 | |||
FQ220741.1 | - | 3 | 283 | |||
FQ225130.1 | - | 3 | 291 | |||
FQ235131.1 | - | 3 | 283 |