Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007654.1 | NP_001007655.1 | 5 | 433 | ||
GenBank mRNA | BC082019.1 | - | 5 | 433 | 7129329 |