Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100886.1 | NP_001094356.1 | 10 | 1266 | ||
XM_006240868.2 | XP_006240930.1 | 9 | 1336 | |||
XM_006240869.2 | XP_006240931.1 | 10 | 1212 | |||
XM_006240870.2 | XP_006240932.1 | 9 | 1180 | |||
GenBank mRNA | BC100259.1 | - | 10 | 1211 |