Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106770.1 | NP_001100240.1 | 3 | 424 | ||
GenBank mRNA | FQ211295.1 | - | 3 | 417 | ||
FQ211839.1 | - | 3 | 407 | |||
FQ217217.1 | - | 3 | 407 | |||
FQ221492.1 | - | 3 | 411 | |||
FQ224331.1 | - | 3 | 411 | |||
FQ224393.1 | - | 3 | 407 | |||
FQ224740.1 | - | 3 | 409 |