Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106884.1 | NP_001100354.1 | 2 | 444 | ||
XM_006244424.2 | XP_006244486.1 | 2 | 512 | |||
XM_006244425.2 | XP_006244487.1 | 2 | 509 | |||
GenBank mRNA | FQ229837.1 | - | 2 | 341 |