Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249584.2 | XP_006249646.1 | 23 | 6568 | ||
XM_006249585.2 | XP_006249647.1 | 22 | 7085 |