Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Patz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107231.2 | NP_001100701.1 | 4 | 1538 | ||
NM_001277214.1 | NP_001264143.1 | 5 | 1611 | |||
XM_006251292.2 | XP_006251354.1 | 5 | 1698 | |||
GenBank mRNA | CK479676.1 | - | 5 | 417 | ||
DY318319.1 | - | 4 | 342 | |||
EV764152.1 | - | 4 | 370 |