Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113779.1 | NP_001107251.1 | 6 | 769 | ||
GenBank mRNA | BC158789.1 | - | 6 | 769 | 7306904 |