Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267534.1 | NP_001254463.1 | 15 | 1514 | ||
GenBank mRNA | BC083866.1 | - | 15 | 1514 |