Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107783.2 | NP_001101253.2 | 4 | 318 | ||
XM_006235108.2 | XP_006235170.1 | 4 | 471 | |||
XM_006235109.2 | XP_006235171.1 | 4 | 471 | |||
XM_006235112.2 | XP_006235174.1 | 4 | 453 | |||
XM_006235113.2 | XP_006235175.1 | 4 | 453 | |||
XM_008762263.1 | XP_008760485.1 | 5 | 359 | |||
XM_008762264.1 | XP_008760486.1 | 4 | 283 | |||
XM_008762265.1 | XP_008760487.1 | 4 | 1047 | |||
XM_008762267.1 | XP_008760489.1 | 4 | 453 | |||
XM_008762268.1 | XP_008760490.1 | 4 | 453 |