Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107865.1 | NP_001101335.1 | 12 | 1585 | ||
XM_006236673.2 | XP_006236735.1 | 13 | 1894 | |||
XM_006236674.2 | XP_006236736.1 | 13 | 1891 | |||
XM_006236675.2 | XP_006236737.1 | 12 | 1852 |