Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107896.1 | NP_001101366.1 | 19, 20 | 2211 | ||
XM_008763382.1 | XP_008761604.1 | 25, 26 | 3325 | |||
XM_008763383.1 | XP_008761605.1 | 24, 25 | 2946 | |||
XM_008763384.1 | XP_008761606.1 | 23, 24 | 3176 | |||
XM_008763385.1 | XP_008761607.1 | 23, 24 | 3173 | |||
XM_008763386.1 | XP_008761608.1 | 23, 24 | 3173 | |||
XM_008763387.1 | XP_008761609.1 | 22, 23 | 3143 | |||
XM_008763388.1 | XP_008761610.1 | 23, 24 | 3113 | |||
XM_008763389.1 | XP_008761611.1 | 22, 23 | 3080 | |||
XM_008763390.1 | XP_008761612.1 | 21, 22 | 3050 | |||
XM_008763391.1 | XP_008761613.1 | 20, 21 | 3017 | |||
XM_008763392.1 | XP_008761614.1 | 22, 23 | 3140 | |||
XM_008763393.1 | XP_008761615.1 | 21, 22 | 3047 |