Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015018.1 | NP_001015018.1 | 2 | 818 | ||
GenBank mRNA | BC092629.1 | - | 2 | 818 |