Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fem1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025706.1 | NP_001020877.1 | 1 | 1074 | ||
GenBank mRNA | BC097490.1 | - | 1 | 1074 | 7460827 |