Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145755.2 | NP_001139227.2 | 9, 10 | 2840 | ||
XM_006246755.2 | XP_006246817.1 | 10, 11 | 3128 | |||
GenBank mRNA | FJ665183.1 | - | 9, 10 | 2831 | ||
HM060628.1 | - | 9, 10 | 2840 | |||
HM060629.1 | - | 9, 10 | 2840 | |||
HM060630.1 | - | 9, 10 | 2824 | |||
HM060631.1 | - | 9, 10 | 2840 | |||
HM060632.1 | - | 9, 10 | 2840 | |||
HM060633.1 | - | 9, 10 | 2824 | |||
HM060634.1 | - | 9, 10 | 2840 | |||
HM060635.1 | - | 9, 10 | 2824 | |||
HM060636.1 | - | 9, 10 | 2824 | |||
HM060637.1 | - | 9, 10 | 2840 | |||
HM060638.1 | - | 9, 10 | 2824 | |||
HM060639.1 | - | 9, 10 | 2824 |