Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199403.1 | NP_955435.1 | 21 | 3040 | ||
XM_006250164.2 | XP_006250226.1 | 21 | 3495 | |||
XM_006250166.2 | XP_006250228.1 | 20 | 3474 | |||
XM_008769657.1 | XP_008767879.1 | 21 | 3507 | |||
XM_008769658.1 | XP_008767880.1 | 20 | 3486 | |||
XM_008769659.1 | XP_008767881.1 | 21 | 3507 | |||
GenBank mRNA | AJ421447.1 | - | 21 | 3040 |