Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108377.2 | NP_001101847.2 | 3, 4 | 569 | ||
GenBank mRNA | BC166766.1 | - | 3, 4 | 542 | ||
FQ226789.1 | - | 3, 4 | 567 |