Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191853.1 | NP_001178782.1 | 3 | 204 | ||
XM_006234653.2 | XP_006234715.1 | 3 | 248 | |||
XM_006234654.2 | XP_006234716.1 | 3 | 237 |