Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134957.1 | NP_001128429.1 | 15 | 3490 | ||
XM_006241712.2 | XP_006241774.1 | 16 | 5700 | |||
XM_006241713.2 | XP_006241775.1 | 15 | 5676 | |||
XM_006241714.2 | XP_006241776.1 | 16 | 5667 | |||
XM_008765515.1 | XP_008763737.1 | 16 | 3678 |