Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766019.1 | XP_008764241.1 | 3 | 452 | ||
XM_008766020.1 | XP_008764242.1 | 3 | 452 |