Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_343965.6 | XP_343966.4 | 2 | 1066 | ||
GenBank mRNA | AY863055.1 | - | 1 | 14 |