Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017468.2 | NP_001017468.1 | 2 | 254 | ||
GenBank mRNA | AB113963.1 | - | 2 | 244 |