Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vpreb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108930.1 | NP_001102400.1 | 1 | 65 | ||
XM_006256322.1 | XP_006256384.1 | 1 | 358 |
Gene Symbol: LOC103694871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773820.1 | XP_008772042.1 | 1 | 358 |