Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009480.1 | NP_001009480.1 | 3 | 174 | ||
GenBank mRNA | BC086954.1 | - | 3 | 174 | 7368408 | |
FQ217127.1 | - | 3 | 219 | |||
FQ217158.1 | - | 3 | 214 | |||
FQ217230.1 | - | 3 | 211 | |||
FQ217709.1 | - | 3 | 218 | |||
FQ217710.1 | - | 3 | 204 | |||
FQ217725.1 | - | 3 | 215 | |||
FQ218028.1 | - | 3 | 204 | |||
FQ221521.1 | - | 3 | 218 | |||
FQ221648.1 | - | 3 | 218 | |||
FQ221867.1 | - | 3 | 207 | |||
FQ222083.1 | - | 3 | 211 | |||
FQ223801.1 | - | 3 | 204 | |||
FQ223893.1 | - | 3 | 218 | |||
FQ223943.1 | - | 3 | 215 | |||
FQ224007.1 | - | 3 | 216 | |||
FQ224711.1 | - | 3 | 189 |