Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238749.2 | XP_006238811.1 | 5 | 635 | ||
XM_008764088.1 | XP_008762310.1 | 1788 | ||||
XM_008764089.1 | XP_008762311.1 | 1608 | ||||
XM_008764090.1 | XP_008762312.1 | 5 | 822 | |||
XM_345572.8 | XP_345573.5 | 5 | 816 |