Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134781.1 | NP_001128253.1 | 17 | 2569 | ||
XM_006236049.2 | XP_006236111.1 | 17 | 2821 | |||
XM_006236050.2 | XP_006236112.1 | 18 | 3675 | |||
XM_006236051.2 | XP_006236113.1 | 17 | 2271 | |||
XM_008762709.1 | XP_008760931.1 | 17 | 3793 | |||
XM_008762710.1 | XP_008760932.1 | 18 | 3187 | |||
XM_008762711.1 | XP_008760933.1 | 17 | 3793 |