Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1c6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003977.1 | NP_001003977.1 | 5 | 717 | ||
XM_006229110.2 | XP_006229172.1 | 5 | 795 |