Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-M6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008852.1 | NP_001008852.1 | 2 | 109 | ||
XM_008772698.1 | XP_008770920.1 | 2 | 110 | |||
GenBank mRNA | EU091353.1 | - | 1 | 24 | ||
EU106176.1 | - | 1 | 24 |