Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dear | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004448.1 | NP_001004448.1 | 1 | 2931 | ||
GenBank mRNA | AY664492.1 | - | 1 | 2931 |
Gene Symbol: Fbxw7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729089.4 | XP_002729135.3 | 4 | 1127 | ||
XM_003749311.3 | XP_003749359.1 | 4 | 936 | |||
XM_006232554.2 | XP_006232616.2 | 5 | 1033 | |||
XM_006232555.2 | XP_006232617.1 | 4 | 3214 |